Bond Elut Certify Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
A396091C A496091C A3960950
CAAGA396091CEA 1030.49 CAD
CAAGA396091C CAAGA496091C CAAGA3960950
Bond Elut Certify Plates, Agilent Technologies
Microplates Solid Phase Extraction Plates (SPE)
Bond Elut Certify is a mixed-mode phase ideal for extraction of basic drugs from urine and blood.
  • Mixed-mode sorbent bed
  • Broad application range for aqueous extraction
  • Bimodal, nonpolar, and strong cation exchange
The Bond Elut Certify plate utilizes a packed bed consisting of a nonpolar C8 sorbent and a strong cation-exchange (SCX) sorbent. Certify is most commonly used to extract basic (cationic) drugs from urine and blood. It is also very effective for extracting a wide range of compounds from a diverse number of aqueous matrices.