Plate, Round Well, J.T.Baker®
Supplier: Ritter, part of Avantor
43001-00101 43001-00161 43001-01161
43001-0010CS 296.55 CAD
43001-0010 43001-0016 43001-0116
Plate, Round Well, J.T.Baker®
Microplates Deep Well Microplates
Made of virgin PP to minimise extractables.
  • Raised rim to prevent cross contamination and enable a better heat-welded seal
  • Also available rimless to allow insertion of vials, or when adhesive seals are used
  • Round bottom for optimal mixing and recovery
  • Suitable for use with automated sample handling systems
  • Can be stored at –80 °C
J.T.Baker Plate Round well plus with air escapes and raised rims for unfreezing samples.