Multi-position Submersible Magnetic Stirrers, Crystal Industries
Supplier: CRYSTAL TECHNOLOGY & INDUSTRIES
MS-04BSUA MS-06BSUA MS-15BSUA
76516-660EA 1447.08 CAD
76516-660 76516-662 76516-664
Multi-position Submersible Magnetic Stirrers, Crystal Industries
Stirrers Magnetic Stirrers, Multi-position
These multi-position submersible magnetic stirrers feature a space saving design, ideal for stirring multiple size beakers or flasks simultaneously and have been constructed to operate safely in a water bath or CO₂ incubator.
  • Stir a variety of beaker or flask sizes simultaneously in a compact space
  • Waterproof body for submersible use in water baths
  • Five stirring levels provide excellent mixing capability
  • Sealed stainless steel body construction protects against intrusion when used in a CO₂ incubator
  • 2-years replacement warranty
Multi-positionMore Product Information