Multi-position Hotplate Stirrers, Crystal Industries
Supplier: CRYSTAL TECHNOLOGY & INDUSTRIES
MS-05HA MS-10HA MS-15HA
76516-668EA 1218.6 CAD
76516-668 76516-670 76516-672
Multi-position Hotplate Stirrers, Crystal Industries
Stirrers Magnetic Hotplate Stirrers, Multi-position
These multi-position hotplate stirrers feature a space saving design, ideal for applications that require both stirring and heating.
  • Fast heating speed and excellent temperature uniformity
  • Heat and stir a variety of different sized beakers or flasks simultaneously
  • Exterior is aluminum alloy for high temperature resistance, corrosion protection and excellent insulation properties
  • Two-year replacement warranty
Multi-position hotplate stirrers are simple to use and save valuable bench space. Simply set the speed and temperature for the application. IdealMore Product Information