HPLC Starter Kits, Supply V2.0, SCAT
Supplier: SCAT AMERICAS, INC.
399200 399201 399202
76503-980PK 694.24 CAD
76503-980 76503-982 76503-984
HPLC Starter Kits, Supply V2.0, SCAT
Closures Safety Caps and Safety Waste Caps
Starter kits consisting of SCAT SafetyCaps, blind plugs, fittings, and air valves for the supply of HPLC systems.
  • Price advantage compared to ordering piece by piece
  • Suitable for all current HPLC systems
  • Unused connectors can be closed by the blind screws delivered with the set, which makes the system universally applicable, even when fewer connectors are needed
Initial removal equipment that can be used with all common HPLC systems, for the safe extraction of solvents.
Ordering information: All set components can be reordered separately.