Safety Funnels with Lid 'MARCO' V2.0, Electrically Conductive, SCAT Europe
Supplier: SCAT AMERICAS, INC.
306980 317637 317651 318630 318960 318961 318962 318964 318980 318981 318983 318984 318985
76503-740EA 259.89 CAD
76503-740 76503-880 76503-902 76503-904 76503-918 76503-920 76503-922 76503-924 76503-926 76503-928 76503-930 76503-932 76503-934
Safety Funnels with Lid 'MARCO' V2.0, Electrically Conductive, SCAT Europe
Closures Safety Caps and Safety Waste Caps
S.C.A.T. 'MARCO' V2.0 safety funnels can be closed safely and prevent danger of ignition.
  • 'MARCO', V2.0 safety lid funnel is a modern lid funnel made of high-quality HDPE with extremely high resistance to acids, alkalis and many chemicals
  • Integrated gripping edge on the funnel lid increases the grip without having to touch the possibly contoured lid
  • Innovative funnel lid with integrated sealing ring closes the funnel edge and prevents explosive or health-endangering gases escaping
  • Sturdy hinge guarantees the closing mechanism has a long service life
More Product Information