IceRack™ Crushed Ice MicroTube Workstations, MTC Bio
Supplier: MTC BIO, INC.
R8012 R8013 R8014
470336-514CS 54.05 CAD
470336-514 470336-516 470336-518
IceRack™ Crushed Ice MicroTube Workstations, MTC Bio
Racks Tube Racks
These tube boxes have a removable white tube tray that can be kept filled with crushed ice or other temperature controlling materials.
  • Designed for crushed ice
  • Polypropylene, –80 to +135 °C
  • Detachable 'lift-off' lids
  • Economical
  • For screw cap and snap cap tubes
They also have a translucent cover. Temperature sensitive samples and assays in microtubes can be processed with an extra degree of temperature stability. Suitable for use with screw cap or snap cap microtubes from 0.5 to 1.5/2.0 ml.
Delivery information: Supplied six boxes per pack.