Deep Well Storage Plates, 96 Round Well, Irish Life Sciences
Supplier: Irish Life Sciences
1.2R96-009U 1.2R96-009US 1.2R96-010UNR 2R96-007U 2R96-007US R96-300V 1R96-304ULP 1R96-322VLP 1.1R96-014ULP 1.1R96-014ULPS
76509-398CS 628.71 CAD
76509-398 76509-400 76509-402 76509-404 76509-406 76509-408 76509-410 76509-412 76509-414 76509-416
Deep Well Storage Plates, 96 Round Well, Irish Life Sciences
Microplates Deep Well Microplates
Round well plates made from medical grade polypropylene. Ideal for storage and screening applications.
  • U and V bottom well format for optimal sample recovery
  • Alphanumeric well coding
  • High temperature and chemical resistance (–80 to +121 °C)
  • Autoclavable
  • RNase, DNase Free
  • Stackable for use in plate and automation hotels
This plate has been developed precisely to conform to the footprint dimensions defined by SLAS/ANSI standards. This guarantees its compatibility with both manual and automated sample handling systems, readers, and microplate washers.
Certifications: Manufactured in ISO Class 8 Cleanroom.
Plate conforms to SLAS/ANSI standards.