Eagle Yellow Infectious Linen Bags
Supplier: Justrite
1692 1694
76520-560PK 375.72 CAD
76520-560 76520-564
Eagle Yellow Infectious Linen Bags
Bags Waste Disposal Bags
Minimize waste by segregating infectious linens.
  • Yellow In color
  • Linear low density polyethylene (LLDPE)
  • 1.2 Mil thickness
  • A high level of flexibility.
  • Shear resistance can be attained without leaks
  • Meets ASTM D 1709-01 and ASTM D 1922-00a specifications
Minimize waste by segregating infectious linen to be laundered. The 33 gal. capacity bag is designed to fit Eagle 1601 series drums. Once the bag is full and ready for laundering, simply seal the bag in place and close the drum using its lid and lever-lock.
Yellow color bag is easily identifiable as infectious linen, drums can be yellow, black, or blue. Printing includes English, Spanish and French translation along with the international recognizable biohazard symbol.
Using Linear Low Density Polyethylene (LLDPE) material at 30.4 µm thickness, a high level of flexibility and shear resistance can be attained without leaks. A tubular construction, single side seal and star seal bottom provide a stable, leak proof bag.
Packaging: Sold in packs of 100.