VWR® Metal Dissecting Scalpel
Supplier: VWR
VWR®
11647-349 11647-350 11647-351
76457-516EA 21.09 CAD
76457-516 76457-518 76457-520
VWR® Metal Dissecting Scalpel
Scalpels and Scalpel Assemblies Scalpels
VWR® Standard Metal Scalpel is perfect for dissecting applications.
  • Easy to clean and sterilize
  • Reusable
  • Smooth, antiglare finish handle
  • Useful in educational, research or healthcare setting
  • Fully autoclavable
Nickel-plated Scalpels.
Certifications: ISO, GMP