VWR® Stainless Steel Scalpel Blades
Supplier: VWR
VWR®
11647-332 11647-327 11647-314 11647-330 11647-313 11647-326 11647-312 11647-329 11647-328 11647-331
76457-480PK 146.14 CAD
76457-480 76457-470 76457-446 76457-442 76457-476 76457-444 76457-478 76457-482 76457-472 76457-474
VWR® Stainless Steel Scalpel Blades
Scalpels and Scalpel Assemblies Scalpel Blades
VWR® Stainless Steel Surgical Scalpel Blades are made from sharp, long lasting surgical stainless steel.
  • Exceptionally strong and unifoirmly sharpened to maximum acuity
  • Surgical grade, stainless-steel blades
  • Individually sealed with peelable packaging
  • Sterilized with gamma radiation
Sterile. 100 per pack.
Certifications: ISO, GMP