VWR® Jar Forceps with Serrated Tips
Supplier: VWR International
76457-410 76457-412 76457-414
76457-410EA 31.29 CAD
76457-410 76457-412 76457-414
VWR® Jar Forceps with Serrated Tips
Forceps
These high-quality thumb forceps remove specimens from deep bottles or jars.
  • Manufactured from AISI 400 series stainless steel
  • High polish, mirror finish
  • Autoclavable and corrosion-resistant
For classroom dissecting, field-study, removal of small embedded objects and crafts.
Certifications: ISO, GMP