VWR® General Purpose Dissecting Scissors
Supplier: VWR International
76457-374 76457-376 76457-378 76457-380 76457-382 76457-384 76457-386 76457-388 76457-390 76457-392 76457-394 76457-396 76457-398 76457-400 76457-402 76457-404 76457-406 76457-408
76457-374EA 37.63 CAD
76457-374 76457-376 76457-378 76457-380 76457-382 76457-384 76457-386 76457-388 76457-390 76457-392 76457-394 76457-396 76457-398 76457-400 76457-402 76457-404 76457-406 76457-408
VWR® General Purpose Dissecting Scissors
Scissors
These high-quality dissecting scissors are primarily used for general and multipurpose cutting, suture removal and dissection applications.
  • Large rings for comfort, user control and stability
  • Manufactured from AISI 420 stainless steel
  • High polish, mirror finish
  • Autoclavable and corrosion-resistant
Product conforms to international quality standards.
Certifications: ISO, GMP
Order Now
SPECIFICATIONS
Environmentally Preferable