Spillage Trays, S.C.A.T.
Supplier: SCAT AMERICAS, INC.
108981 117983 117984 117985 117986
76503-592EA 544.42 CAD
76503-592 76503-628 76503-630 76503-632 76503-634
Spillage Trays, S.C.A.T.
Trays/Dishes/Pots Spillage Trays/Pans
Made of HDPE, white or black (electrically conductive). Containers for all purposes.
  • Electrically conductive available
  • Available with base insert; the canisters always stay dry
  • Anti-static mat and ground cable optionally available
Suitable for a wide range of canisters. Collect liquid in case of a leakage. The SCAT collection trays prevent the uncontrolled spread of hazardous liquids in the event of damage to a disposal container. The HDPE-EC conductive tray also prevents the build-up of static charge, which prevents the ignition of explosive substances.
Order Now
SPECIFICATIONS
Environmentally Preferable