VWR® Post-Mortem Autopsy Scalpel Blades
Supplier: VWR International
76457-426 76457-440
76457-426PK 185.66 CAD
76457-426 76457-440
VWR® Post-Mortem Autopsy Scalpel Blades
Scalpels and Scalpel Assemblies Scalpel Blades
These high-quality Post Mortem Autopsy Scalpel Blades are made from surgical carbon steel blades and sharpened to perfection for a super clean cut.
  • Thicker blades reduce breakage
  • Surgical-quality blades
  • Individually sealed and peelable packaging
  • Sterilized with gamma radiation
For post-mortem, autopsy and research use. Fits VWR® Post Mortem Scalpel Knives.
Certifications: ISO, GMP