VWR® Scalpel Knife Handle
Supplier: VWR International
76457-416 76457-418 76457-420
76457-416EA 26.63 CAD
76457-416 76457-418 76457-420
VWR® Scalpel Knife Handle
Scalpels and Scalpel Assemblies Scalpel Handles
These high-quality scalpel knife handle fits the VWR® 10 and 20 series scalpel blades.
  • Secure blade fitment
  • Autoclavable/sterilizable
  • Ribbed/grooved
For classroom dissection, field-study, removal of small embedded objects and crafts.
Fits VWR® carbon and stainless steel scalpel blades.
Certifications: ISO, GMP