VWR® Dissecting Scissors with Open Shanks
Supplier: VWR
VWR®
11647-277 11647-276
76457-370EA 16.77 CAD
76457-370 76457-372
VWR® Dissecting Scissors with Open Shanks
Scissors
These high-quality dissecting scissors are primarily used for general, multipurpose cutting and dissection applications.
  • Large rings for comfort, user control and stability
  • Manufactured from AISI 420 stainless steel
  • High polish, mirror finish
  • Autoclavable and corrosion-resistant
Product conforms to international quality standards.
Certifications: ISO, GMP