Eisco LabGlass™ Low Form Glass Beakers
Supplier: Wards
CH0126L CH0126A CH0126B CH0126C CH0126D CH0126E CH0126F CH0126G CH0126H CH0126I CH0126J CH0126J8 CH0126K CH0126M
470335-848EA 36.2 CAD
470335-848 470335-850 470335-852 470335-854 470335-856 470335-858 470335-860 470335-862 470335-864 470335-866 470335-868 470335-870 470335-872 470335-874
Eisco LabGlass™ Low Form Glass Beakers
Beakers
High quality borosilicate 3.3 glass beakers with a spout for easy pouring. Designed for general use in a wide variety of laboratories.
  • Made from high quality borosilicate 3.3 glass
  • Low form design with spout for easy pouring
  • Superior chemical durability
  • Low temperature gradient, can withstand thermal shock
  • Uniform wall thickness for superior durability
  • Autoclavable at 121 °C
  • Graduated per DIN 12231, ISO 3819
Eisco low form beakers are made of high-quality Borosilicate 3.3 glass. These durable glass beakers have a low temperature gradient and can withstand high temperatures as well as thermal shocks. They are microwaveable, autoclavable, and have superior chemical durability. Eisco Labs Borosilicate glass has the following heat tolerances: Strain Point –515 °C | Annealing Point 565 °C | Softening Point 820 °C.
Eisco Labs proudly manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Certifications: DIN 12231, ISO 3819