VWR® Critical Environment Swab, Mini Flexible Foam Head
Supplier: VWR International
76478-846
76478-846BX 156.77 CAD
76478-846
VWR® Critical Environment Swab, Mini Flexible Foam Head
Swabs Swabs for Clinical Use
Swab features 100 ppi open cell polyurethane flexible foam head with 100% virgin polypropylene handle.
  • Polyurethane foam swab head
  • Polypropylene handle
  • Flexible, mini foam head
Swab head is thermally bonded to the handle without using any adhesive.