PIPETMAX® Pipette Heads, Gilson
Supplier: GILSON, INC.
FC10022 FC10021 FC10010 FC10023
76518-582EA 4285.75 CAD
76518-582 76518-584 76518-586 76518-588
PIPETMAX® Pipette Heads, Gilson
Automated Liquid Handling Systems
PIPETMAX heads are specially designed for the PIPETMAX system.
The PIPETMAX® offers 8-channel pipette heads with the system. Choose from three options, pipetting volumes between 1 and 10 µl, pipetting volumes between 20 and 200 µl and fixed 1 ml volumes.
Accessories information: PIPETMAX D1200 Tip Reload Box and 4×1200 Tip Ejector pairs with the MAX4×1200 Pipette Head.
Order Now
SPECIFICATIONS
Environmentally Preferable