SPEC Disk SPE, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
A5321130 A5320320 A5322020 A532DAU A5022570 A5321120 A5321920
CAA5321130PK 802.54 CAD
CAA5321130 CAA5320320 CAA5322020 CAA532DAU CAAGA5022570 CAAGA5321120 CAAGA5321020 CAAGA5321920
SPEC Disk SPE, Agilent Technologies
Chromatography Columns
SPEC disk SPE delivers superior flow characteristics and trouble-free automation.
  • No loose sorbent means no channeling and better reproducibility
  • Uniform flow and extraction properties offer robust performance
  • Low elution volume affords excellent concentration of the analyte, improving sensitivity
SPEC disk SPE is a product providing faster sample processing, lower costs, and high-quality results. SPEC is manufactured with ultraclean polypropylene plastic and contains a monolithic sorbent bed with bonded silica infused on a glass fiber disk. For some applications, the elution volume is very small, eliminating the dry down and reconstitution steps.
Integrated filtration protects against clogging. SPEC is available in cartridge, 96-well plate, and disk formats in a wide range of chemistries for many applications.