Empty Cartridges and Empty Cartridges with Pre-installed Frits, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
12131010 12131011 12131012 12131016 12131018 12131008 12131015 12131017
CA12131010PK 200 CAD
CA12131010 CA12131011 CA12131012 CA12131016 CA12131018 CAAG12131008 CAAG12131015 CAAG12131017
Empty Cartridges and Empty Cartridges with Pre-installed Frits, Agilent Technologies
Chromatography Columns
Empty cartridges can be used to increase Bond Elut cartridge capacity or for custom SPE packing.
  • Made with high purity polypropylene for cleaner extracts
  • Uniform batch-to-batch size for consistent performance
  • Economical for everyday use
A variety of empty reservoirs is available to increase the sample capacity of Bond Elut extraction cartridges. These tubes can also be used to pack custom SPE cartridges with bulk Bondesil sorbents or others and are available from 1 to 60 ml.
Ordering information: Cartridges come with pre-installed frits, or 20 µm frits can be ordered separately.