Dissolution Sampling Cannulas
Supplier: Agilent
CrossLab
12-3200 12-3215 17-3306 27-6500 12-3222 12-3230 17-3307 17-3300 17-3306 12-3219 17-3305 12-3215 12-3221 17-3307 12-3224 27-6500 12-3210 12-3219 12-3200 12-3216 12-3223 12-3216 17-3300 12-3221 17-3305 12-3224 12-3210 12-3222 12-3223
CAAG12-3224EA 115.12 CAD
AG12-3215 CAAG12-3224 CAAG12-3223 AG12-3216 AG12-3219 AG12-3230 AG12-3210 CAAG12-3200 CAAG12-3222 CAAG12-3221 AG17-3300 AG17-3306 AG17-3305 AG27-6500 AG17-3307 CA17-3306 CAAG12-3215 CAAG12-3216 CAAG12-3219 AG12-3200 AG12-3222 AG12-3221 AG12-3224 CAAG12-3210 AG12-3223 CAAG17-3300 CAAG17-3307 CAAG17-3305 CAAG27-6500
Dissolution Sampling Cannulas
Dissolution Apparatus
Available in two different lengths based on the volume of media; best used with full flow filters.
  • For manual sampling, the Agilent cannula luer lock assembly is comprised of a bent cannula and a sampling syringe that are best used with the full flow filters
  • The sampling cannulas are available two in different lengths based on the volume of media: 500 or 900 ml
  • An adjustable gauge can be included with this assembly to ensure repeatable positioning for each sample
The Agilent cannula luer lock assembly is comprised of a bent cannula and a sampling syringe that are best used with the full flow filters. The sampling cannulas are available in two different lengths based on the volume of media being sampled from.