CrossLab Tubing and Fittings for 8453/8454 UV Systems
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5022-2154 5022-2156 5022-2157 5041-2166 5041-2184 5041-2185 5041-2191 5042-1330 5042-1333 5021-1870 5022-2158 5042-1331 5042-1337 5041-2199
CA5022-2154PK 77.55 CAD
CA5022-2154 CA5022-2156 CA5022-2157 CA5041-2166 CA5041-2184 CA5041-2185 CA5041-2191 CA5042-1330 CA5042-1333 CAAG5021-1870 CAAG5022-2158 CAAG5042-1331 CAAG5042-1337 AG5041-2199
CrossLab Tubing and Fittings for 8453/8454 UV Systems
Chromatography Supplies Chromatography Spare Parts
Tubing and Fittings are designed for 8453 and 8454 UV-Vis Instruments and Accessories.
  • These quality supplies provide the perfect fit with your Agilent instrument
  • Why risk compromising your analytical results with anything less than genuine Agilent consumables
Agilent offers replacement tubing, fittings and kits for the 8453 and 8454 UV-Vis spectrophotometer systems. These allow you to replace peristaltic pump tubing, inlet/outlet connections for flow cells and dissolution test systems to help you keep your system operating at peak performance.