CrossLab Spray Chamber End Caps for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G3280-60008 G8400-40200
CAG3280-60008EA 545.91 CAD
CAG3280-60008 CAAGG840040200
CrossLab Spray Chamber End Caps for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Spray chamber end cap, nebulizer retaining nut and other supplies for ICP-MS Scott spray chamber.
  • Optional inert PFA Scott type spray chamber provides excellent sensitivity, stability and washout
  • PFA end cap is O-ring free and features a 6 mm nebulizer port and a make up gas port
  • Manufactured from the highest purity grade PFA resin for the lowest blank contribution, ensuring it is suitable for semiconductor applications
The standard quartz sample introduction kit for Agilent ICP-MS instruments features a quartz Scott-type spray chamber offeringMore Product Information