CrossLab Split Liners for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
18740-60840 18740-80190 5183-4697 5183-4699 5183-4713 5183-4700 5183-4702 19251-60540 5183-4647 5183-4711 5183-4712 210-4004-5 5183-4691 5183-4692 5183-4701 210-4022-5 5190-2275
CA18740-60840EA 112.24 CAD
CA18740-60840 CA18740-80190 CA5183-4697 CA5183-4699 CA5183-4713 CA5183-4700 CA5183-4702 CA76508-028 CA76508-160 CA76508-162 CA76508-164 CA76514-580 CA76514-898 CA76514-900 CA76514-904 CA210-4022-5 CAAG5190-2275
CrossLab Split Liners for GC, Agilent Technologies
Chromatography Supplies Chromatography Inlet Liners
Agilent split liners are made to strict dimensional specifications for optimal inlet performance.
  • Feature Agilent’s exclusive deactivation;Agilent recommends part number 5183-4647 as the top split liner choice
Split injections are intended for use with a split/splitless injection port in gas chromatography analysis. Split liners tend to have a slightly smaller outer diameter to allow for gas flow. For ease of use and reproducibility, the liners have a positioning bead, a restriction to secure the position of the glass wool, and a feature toMore Product Information