CrossLab Semi-Microcells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5061-3391 5063-6559 5063-6561 6610001800 6610012700 6610015400 6610021100 5063-6558 6610019600 6610019800
CA5061-3391EA 796.7 CAD
CA5061-3391 CA5063-6559 CA5063-6561 CAAG6610001800 CAAG6610012700 CAAG6610015400 CAAG6610021100 CAAG5063-6558 CAAG6610019600 CAAG6610019800
CrossLab Semi-Microcells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
Cuvettes Spectrophotometer Cuvettes
Agilent offers a wide range of semi-micro rectangular cuvettes with path lengths from 1 to 10 mm.
  • Semi-micro cells feature non transmitting, self masking black walls which improve sensitivity by eliminating stray light from measurements
  • Fill volume reduced to approximately 40% of that used with a standard 10 mm path length rectangular cell
  • Exceptional accuracy in terms of optical path length and parallelism
  • Unique surface flatness of the optical windows
  • High temperature stability
  • High chemical resistance
  • Outstanding measurement reproducibility
  • Guaranteed
More Product Information