Cross-linked and bonded μSIL capillaries
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
194-8111 196-1002 199-2602
CA194-8111PK 638.37 CAD
CA194-8111 CAAG196-1002 CAAG199-2602
Cross-linked and bonded μSIL capillaries
Electrophoresis Systems Capillary Electrophoresis Systems
µSIL-FC and µSIL-DNA capillaries are chemically inert, hydrophobic, and stable from pH 2.5 to 10.0.
  • µSIL-DNA capillaries coated with an FC polymer
  • 75 µm id to accommodate the viscosity of entangled polymer solutions
  • All µSIL capillaries are batch tested to ensure the highest performance and reproducibility
These capillaries are a must-have for cIEF, protein, peptide and carbohydrate separations, as well as replaceable gel CE applications such as oligonucleotides, DNA fragments, and PCR product separations. The µSIL capillaries are permanentlyMore Product Information