CrossLab Pre-Optics Cone and Windows for ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
3610007300 G8010-68014 G8010-68015 G8010-68109 1510220200 G8010-68018
CA3610007300EA 1094.44 CAD
CA3610007300 CAG8010-68014 CAG8010-68015 CAG8010-68109 CAAG1510220200 CAAGG801068018
CrossLab Pre-Optics Cone and Windows for ICP-OES, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Pre-optics cones and axial/radial windows for Agilent ICP-OES.
  • Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
  • Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise
Agilent supplies and accessories enable you to maintain the superior performance of your ICP-OES. Agilent ICP-OES pre-optics cones and axial/radial windows are available for Agilent 5000 series ICP-OES and legacy 700 series, Vista, and Liberty systems.