CrossLab Nebulizer Components for Mark 7 Spray Chamber, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
1510226700 1510226800 9910093000 310046600 810133400 810134300 4610017400 810134400 1610117390
CA1510226700EA 24.69 CAD
CA1510226700 CA1510226800 CA9910093000 CA310046600 CA810133400 CA810134300 CAAG4610017400 CAAG810134400 CA1610117390
CrossLab Nebulizer Components for Mark 7 Spray Chamber, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
The nebulizer used in the Mark 7 atomization system is adjustable for peak performance with samples.
  • The nebulizer is integrated into the nebulizer block of the spray chamber assembly
  • The user can adjust the nebulizer uptake rate from 0 to approximately 8 ml/min. giving flexibility to handle most solution types
  • Utilizing the high vacuum setting greatly reduces the dependence of the uptake rate on liquid level in the sample container
  • The nebulizer is fully serviceable and can be easily maintained or components replaced by the user
  • Use organic resistant O-
More Product Information