CrossLab Self-Tightening Column Nuts for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G3440-81011 G3440-81013 G3440-81012
CAG3440-81011EA 176.46 CAD
CAG3440-81011 CAG3440-81013 CAG3440-81012
CrossLab Self-Tightening Column Nuts for GC, Agilent Technologies
Fittings and Connectors
Our unique, stainless steel, self-tightening GC column nuts deliver tight connections with no expensive upgrades or adaptors needed.
  • No retightening equals less wated time
  • Finger-tight design lets anyone make quality, consistent connections without tools
  • Low-torque seal prevents ferrules from sticking or crumbling
  • Lower column bleed means longer column life
  • Locking collar keeps column in place during installation to ensure consistent and reproducible results
This unique, stainless steel GC column nut delivers a tight connection – without expensiveMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable