CrossLab Uptake PFA Tubing for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G1820-65105 G1820-65478 G1820-65526 G1820-65527 G1820-65531 G1833-65572 G1833-65573
CAG1820-65105EA 138.66 CAD
CAG1820-65105 CAG1820-65478 CAAGG182065526 CAAGG182065527 CAAGG182065531 CAAGG183365572 CAAGG183365573
CrossLab Uptake PFA Tubing for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
PFA sample uptake tubing with different Int.Ø available to suit your ICP-MS application.
  • PFA tubing is used to introduce sample together with the peristaltic pump tubing to the nebulizer sample inlet
  • It provides chemical resistance, inertness and cleanliness
PFA tubing is used to introduce sample to the peristaltic pump tubing for the sample inlet, and to connect the outlet of this tubing to sample line connector block. This connection grounds the sample introduction tubing, eliminating any build up of electrostatic charge, that could lead to signal instability.
PFA tubing of different Int.Ø's are available so you can select the optimum tubing for your application. Tubing with a PEEK guide to ensure the sample tubing stays in the solution is also available.