Sample Tubes and Vials, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
00000001600L 190047900 190049700 190065200 3710051100 5043-0064
CA00000001600LPK 469.69 CAD
CA00000001600L CA190047900 CA190049700 CA190065200 CA3710051100 CA5043-0064
Sample Tubes and Vials, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Sample tubes and vials for SPS 4, SPS 3, and ASX 500 autosamplers for AAS, ICP-OES, MP-AES and ICP-MS.
  • Most tubes made from autoclavable PP
  • Available in different sizes (Ext.Ø, height and volume) to give you flexibility
  • Maximum tube height of 150 mm
A number of autosamplers have been used with Agilent AA, ICP-OES, MP-AES and ICP-MS instruments, depending on the specific model. This includes the Agilent SPS 4 used with current instruments, and the SPS 3 or ASX 500 Series used with earlier model instruments.
Each autosampler automates theMore Product Information