CrossLab Quartz Spray Chambers for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G3280-80008 G8400-67150 G3280-80503
CAG3280-80008EA 1007.14 CAD
CAG3280-80008 CAG8400-67150 CAAGG328080503
CrossLab Quartz Spray Chambers for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Quartz spray chamber and supplies for Agilent ICP-MS, including for use with HMI or UHMI.
  • Agilent Quartz spray chambers for use with ICP-MS instrument temperature control via MassHunter software will ensure your system performs to its full capacity every time
  • Double pass design ensures smaller tertiary aerosol droplet size for better plasma stability, precision (lower % RSD in results) and enhanced matrix tolerance leading to robust methods
  • Ultra high matrix introduction (UHMI) spray chamber for the 79/8900 increases matrix tolerance up to 25% total
More Product Information