CrossLab Connector Tubes from Spray Chamber to Torch for ICP-MS, Agilent Technologies
Supplier: Agilent
CrossLab
G3270-80024 G3270-80025 G3270-80025 G3270-80024
CAG3270-80024KT 472.57 CAD
AGG3270-80025 AGG3270-80024 CAG3270-80024 CAG3270-80025
CrossLab Connector Tubes from Spray Chamber to Torch for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Straight transfer tube & connector with dilution port for ICP-MS spray chamber to torch connection.
  • Straight connector tube is for instruments with the UHMI spray chamber (pn G8400-67150) or earlier models that do not have HMI capability
  • The connector tube with dilution port inlet is for instruments with HMI capability, to enable introduction of the dilution gas to the aerosol or for addition of option gas (Oxygen) inot the plasma for organic solvent analysis
  • Standard tube is quartz for the quartz Scott spray chamber and single-piece quartz torch
  • Optional inert PFA transfer tube for use with inert sample introduction kits
A quartz connector or transfer tube is used to join the standard quartz spray chamber to the base of the quartz torch for Agilent ICP-MS instruments. Optionally a PFA equivalent is available for use with inert sample introduction kits. Two different connector tubes are available. A straight transfer tube is used with ICP-MS having UHMI spray chamber (pn G8400-67150) or earlier models that do not have HMI capability. A connector tube with a dilution port inlet fitting is used with instruments that have HMI capabilities, to enable introduction of the dilution gas to the aerosol prior to the torch.