CrossLab Tools and Manuals for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5180-4162 G1833-65404 G1960-20033 G7201-90302 G8400-90000 G8400-90001
CA5180-4162EA 816.07 CAD
CA5180-4162 CAG1833-65404 CAAGG196020033 CAAGG328060910 CAAGG720190302 CAAGG840090000 CAAGG840090001
CrossLab Tools and Manuals for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Tools and manuals for Agilent ICP-MS 7700, 7800, 7900 and 8800, 8900 instruments
  • Find all the tools and support documentation/manuals for Agilent 7700, 7800, and 7900 single quadrupole systems and 8800 and 8900 triple quadrupole ICP-MS systems here
  • We provide interface sampler and skimmer cone removal tools and cleaning aids simplify your routine maintenance tasks
  • Order copies of our ICP-MS maintenance video/DVD for training new users and ensuring you have the most up to date procedures for looking after your Agilent instrument
A complete range ofMore Product Information