CrossLab Skimmer Cones for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G1833-65132 G3280-67041 G8400-67200 G3270-65024 G3280-67060 G8400-67201 G8400-67202 G3280-67063 G3280-67066 G3666-67411 G1820-65050 G1820-65237 G1833-65497 G3270-60106 G3270-60643 G3280-67064 G3280-67065 G3666-67401 G3666-67421 G3666-67511
CAG1833-65132EA 5274.89 CAD
CAG1833-65132 CAG3280-67041 CAG8400-67200 CAG3270-65024 CAG3280-67060 CAG8400-67201 CAG8400-67202 CAAGG328067063 CAAGG328067066 CAAGG366667411 CAAGG182065050 CAAGG182065237 CAAGG183365497 CAAGG327060106 CAAGG327060643 CAAGG328067064 CAAGG328067065 CAAGG366667401 CAAGG366667421 CAAGG366667511
CrossLab Skimmer Cones for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent skimmer cones ensure the performance that single & triple quadrupole ICP-MS systems demand.
  • Nickel skimmer cones are suitable for most common applications and provide the most economical operation
  • Platinum skimmer cones are required for analysis of aggressive acids (especially HF) and when O2/Ar option gas is used for analysis of organic solvents
  • Platinum skimmer with copper base is recommended for the lowest LODs and for higher matrix samples
  • Platinum skimmer with nickel base is recommended for organics analysis
Agilent, nickel or platinum tipped skimmer cones use high-purity materials to minimize background and ensure stability within the plasma. Cones are typically the biggest supplies cost in routine ICP-MS operation. Protect your investment and ensure ease of use by choosing genuine Agilent cones.High-quality interface cones are key to the sensitivity and stability of an ICP-MS. Our skimmer cones are manufactured to stringent specifications and rigorously tested to ensure excellent batch to batch reproducibility. The orifice dimensions and tip geometry are optimized and tightly controlled.