CrossLab Autosampler Valve Supplies for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
0101-1417 0100-1852 1535-4045 5068-0209 5068-0099 5068-0229 5068-0232 5068-0006 5067-4131 5320-0010
CA0101-1417EA 1848.76 CAD
CA0101-1417 CA76514-502 CA1535-4045 CA5068-0209 CA5068-0099 CA5068-0229 CA5068-0232 CAAG5068-0006 CAAG5067-4131 CAAG5320-0010
CrossLab Autosampler Valve Supplies for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
A full range of supplies for autosampler injection valves, including rotor seals and stator faces.
  • The rotor is a highly stressed part of autosamplers that is constantly switched back and forth
  • Rotor material and surface finish govern its durability and lifetime
  • Agilent rotors are tested to ensure the longest possible lifetime
  • Use of non-Agilent valve supplies can lead to shorter lifetime, potential carryover, and leakage
Agilent provides a full range of supplies, including rotor seals and stator faces, for different autosampler injection valves.More Product Information