Splitless Liners for GC
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
18740-80220 210-3003-5 5183-4695 5183-4696 5183-4704 210-3003 5183-4694 5183-4703 5183-4705 5062-3587 5181-3315 5181-3316 5181-8818 5183-4693 5181-3316I 5190-2271 392611924 5190-2270 5183-4706 5183-4707 5183-4708 5190-2272
CA18740-80220EA 67.77 CAD
CA18740-80220 CA210-3003-5 CA5183-4695 CA5183-4696 CA5183-4704 CA210-3003 CA5183-4694 CA5183-4703 CA5183-4705 CA76508-058 CA76514-738 CA76514-740 CA76514-752 CA76514-902 CA5181-3316I CA5190-2271 CAAG392611924 CAAG5190-2270 CAAG5183-4706 CAAG5183-4707 CAAG5183-4708 CAAG5190-2272
Splitless Liners for GC
Chromatography Supplies Chromatography Inlet Liners
We offer a complete line of liners for your Agilent GC splitless inlet, with exclusive deactivation.
  • Feature Agilent’s exclusive deactivation
  • Designed and engineered to meet strict specifications for dimensional accuracy and inertness
  • Outer diameter of splitless liners is ground and polished to exacting dimensional tolerances, ensuring a perfect fit into our inlets and optimal splitless injection
  • We recommend trying a liner without glass wool first
  • Use glass wool as a second choice
Choose a liner without glass wool if your sample contains nonvolatiles or analytes within a wide boiling range. For unknown samples, use a liner without glass wool to avoid loss of active or labile compounds.
Agilent offers a complete portfolio of liners for your Agilent GC splitless inlet. Our exclusive deactivation is important because of the longer sample/liner contact in splitless mode.