Captiva Filter Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
A5960045 A5960002 A5960002K A596002000B A596401000 A5967045 A596002000 A5962012 A5960002B
CAA5960045PK 814.96 CAD
CAA5960045 CAA5960002 CAA5960002K CAA596002000B CAA596401000 CAA5967045 CAAGA596002000 CAAGA5962012 CAAGA5960002B
Captiva Filter Plates, Agilent Technologies
Microplates Filter Plates
Captiva dual-depth filtration media removes precipitated proteins and prevents HPLC column clogging.
  • Standard SPE format; ideal for LC/MS samples
  • Avoid sample transfer problems
  • Protect HPLC columns from clogging to reduce instrument downtime
  • Clean and clear filtrates offer improved sensitivity
  • High analyte recovery with simple robust methods allows faster method development
  • Rigorous quality control eliminates nonspecific binding
  • Fast and reliable processing improves productivity
Captiva filtration plates completely remove precipitated proteins and strongly protect against HPLC column clogging. Captiva 96-well filter plates, available in 0.2 and 0.45 µm pore sizes, are ideal for preparing precipitated protein samples for LC/MS. Captiva 10 and 20 µm glass fiber filter plates are perfect for automated systems and for clarifying particle-laden samples.
Delivery information: Captiva 96-well replacement filtration kit includes 5 Captiva filter plates, 10 DuoSeal 96-well plate seals, 5 Captiva 96-Deep well 1 ml collection plates, 5 Captiva collection plate pierceable covers.