Bond Elut™ 96 Square-Well Plates, 2 ml, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5133008 5133009 5133007
CA5133008PK 611.36 CAD
CA5133008 CA5133009 CAAG5133007
Bond Elut™ 96 Square-Well Plates, 2 ml, Agilent Technologies
Microplates Solid Phase Extraction Plates (SPE)
Bond Elut 96 square-well plates support well-plate SPE applications with larger sample volumes.
  • Enhanced quality for trouble-free high throughput
  • Recommended for 96-well plate applications with larger sample volumes
  • Available with many of our most popular Bond Elut™ sorbents
  • Added flexibility for method development
  • Automation-friendly to free up operator time
  • Compatible with the Agilent positive pressure manifold 96 processor (PPM-96)
Bond Elut™ 96 square-well plates facilitate transfer from cartridge-based methods to 96-well plate automated formats.More Product Information