CrossLab Spray Chamber Components for Atomic Absorption Instruments, Agilent Technologies
Supplier: Agilent
CrossLab
9910093600 6210092090 9910093600 9910063600 6210092090 9910105800 1610118800 1510226600 9910093100 9910063600 1610118800 110637000 1510226000 1510226000 1510225000 1510226500 110637000 1510225000 1510226600 9910093100 1610118400 1610118400 1510226500 9910105800
CAAG6210092090EA 197.65 CAD
CAAG6210092090 CAAG1610118400 CAAG9910063600 CAAG1510226000 CAAG110637000 CA1510225000 CA9910093600 AG9910093100 CAAG1610118800 AG1510226000 CAAG9910093100 AG6210092090 AG9910063600 AG1610118400 AG1510225000 AG9910093600 CA9910105800 AG9910105800 AG1510226600 CA1510226500 AG110637000 CA1510226600 AG1510226500 AG1610118800
CrossLab Spray Chamber Components for Atomic Absorption Instruments, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Mark 7 atomization system for current model Agilent AA instruments.
  • Simple-to-use twist and lock assembly makes for easy cleaning and routine maintenance
  • Flexible operation with the capability to tune performance using the externally adjustable impact bead, fit mixing paddles for better precision, or both
  • Inert construction makes spray chamber suitable for use with aqueous and organic solutions
  • Fit organic resistant O-rings to ensure organic compatibility
The nebulizer, spray chamber and burner, plays an important part in controlling the analytical performance of a flame AA spectrometer. Proper spray chamber design ensures the minimization of carbon build-up, burner blockage and volatilization interferences. The spray chamber must also provide excellent drainage.
The Mark 7 spray chamber features an adjustable impact bead that breaks up aerosol droplets and provides the operator with control over the aerosol concentration. A set of removable twin-head mixing paddles promote mixing and large droplet removal. These features provide a smaller drop size aerosol, reducing burner blockage, signal noise, and improving long term signal stability.