CrossLab ORS (Octopole Reaction System) Supplies for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G3280-67045 G3280-20618 G3280-20633 G3280-20664 G3280-67046 G3666-67045 G3666-67046 G3666-67141 G3666-67142
CAG3280-67045EA 1958.55 CAD
CAG3280-67045 CAAGG183365133 CAAGG328020618 CAAGG328020633 CAAGG328020664 CAAGG328067046 CAAGG366667045 CAAGG366667046 CAAGG366667141 CAAGG366667142
CrossLab ORS (Octopole Reaction System) Supplies for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
ICP-MS Octopole Reaction System (ORS) cell components for Agilent 7700, 7800, 7900, 8800, 8900.
  • The Agilent ORS is an octopole ion guide with excellent ion focusing and ion transmission properties.
  • Interaction between the polyatomic ions and the cell gases ensures removal of interference from the mass spectrum by KED in helium collision mode or mass shift using a reactive cell gas
  • Replacement kits and individual components for the ORS cell for routine, x-lens, or semiconductor, s-lens (with or without axial acceleration), configurations are available
  • Find
More Product Information