CrossLab Burners and Supplies for Agilent Instruments, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
6910010500 210164000 9910053900 210164100 6910013100
CA6910010500EA 8.24 CAD
CA6910010500 CA210164000 CA9910053900 CAAG210164100 CAAG6910013100
CrossLab Burners and Supplies for Agilent Instruments, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Mark 7 burner used with Mark 7 flame atomization system for current Agilent flame AA instruments.
  • Compatible with all current Agilent flame AA instruments and the earlier flame AA instruments with the Mark V and VI spray chambers
  • Provides a more stable flame with improved resistance to salt and carbon build up
  • Improves sample throughput and productivity
The Mark 7 burner used with the Mark 7 flame atomization system was designed to provide a more stable flame with improved resistance to salt and carbon build up. This translates into longer runningMore Product Information