AVS 6/7 Supplies for ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G8494-60002 G8494-60000 G8494-60003 G8495-60000 G8495-60003 G8494-60007
CAG8494-60002EA 605.6 CAD
CAG8494-60002 CAAGG849460000 CAAGG849460003 CAAGG849560000 CAAGG849560003 CAG8494-60007
AVS 6/7 Supplies for ICP-OES, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
AVS 6/7 supplies include spare valve, stator and rotor, rotor seal, connection kits, and drip tray.
  • Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
  • Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise.
AVS 6/7 Advanced Valve System (AVS) supplies are available for the Agilent ICP-OES. Maintain your instrument's productivity with these replacement supplies for the switching valve. The AVS 7 has 7 ports with the 7th port for internal standard addition. When an internal standard is not required a plug can be used in the 7th port.