NoTrax® 830 High-Voltage Switchboard Matting, Justrite®
Supplier: Justrite
830R0275BL 830R0375BL 830R0375BY 830R0475BL
76519-288EA 2000.45 CAD
76519-288 76519-290 76519-292 76519-294
NoTrax® 830 High-Voltage Switchboard Matting, Justrite®
Floor Mats Permanent Floor Mats
Corrugated Switchboard Matting is non-conductive, designed to provide insulation for the worker.
  • Insulated matting engineered to protect workers from electric shock
  • Electrically Insulative Type 2 PVC compound
  • Class 2 Di-electric strength: 30000 volts
  • Proof test of 20000 volts
  • Corrugated top surface - Added traction and easy to sweep clean
Corrugated Switchboard Matting is a non-conductive floor matting made from a PVC compound, designed to provide insulation for the worker. The mat prevents the worker from being grounded, thereby eliminating the possibility of electrical shock. The corrugated saw-tooth top surface provides increased traction and is easy to sweep clean. Switchboard matting is intended for use in dry environments in front of high voltage boxes, electrical fuse boxes, and transformers.