Vi-CELL BLU Single-Use Controls (Vials), Beckman Coulter
Supplier: Beckman Coulter
C09150 C09145 C09148 C09150 C09147 C09149 C09149 C09145 C09147 C09148
CA76409-831PK 916.24 CAD
CA76409-831 76409-833 CA76409-833 76409-835 CA76409-835 76409-837 CA76409-837 76409-839 CA76409-839 76409-831
Vi-CELL BLU Single-Use Controls (Vials), Beckman Coulter
Assays Cellular Assays
This single-use control can be used for QC runs as a daily monitoring tool to ensure the performance of your Vi-CELL BLU. Each vial contains 200 μl. Upon receipt, the controls must be stored at 2 to 8 °C.
Ordering information: Box of 25 vials.