Vacuum Filtration Units, Sartorius
Supplier: Sartorius
16510 16511
CA76517-774EA 218.1 CAD
CA76517-774 CA76517-776
Vacuum Filtration Units, Sartorius
Funnels Membrane Filter Funnels
PC, autoclavable.
For the clarification or sterile filtration of up to about 200 ml of aqueous solutions.
512-3310 is complete with receiver flask of ±250 ml for sub-pressure as well as with slight over-pressure.
512-3311 without receiver, can be used in combination with Combisart manifolds.
For sterile filtrations- the filter holder includes a glass fiber filter that enables sterile venting, thus avoiding contamination of the sterile filtrate.