Desiccator Plates with Small Holes
Supplier: United Scientific Supplies
JSD140 JSD190 JSD230
470336-406EA 57.5 CAD
470336-406 470336-408 470336-410
Desiccator Plates with Small Holes
Desiccators Desiccators, Plates
Plates include a large hole in the center and numerous 5 mm diameter perforations. Glazed on one side.
  • Glaze: No softening occurs up to 1000 °C, and it is extremely white
  • Resistance to chemical action: Resistance to acids and alkalies is excellent, except for hydrofluoric acid
Thermal shock resistance: Average coefficient of expansion from 20 to 200 °C is 3.56×10⁻⁵, gradually increasing to 4.69×10⁻⁵ at 1050 °C. Every batch of porcelain is heat tested.