Diamond® PureFlow™ CA Syringe Filters, Sterile, Globe Scientific
Supplier: Globe Scientific
SF-CA-2213-S SF-CA-2230-S SF-CA-4513-S SF-CA-4530-S
76516-614BX 96.8 CAD
76516-614 76516-616 76516-618 76516-620
Diamond® PureFlow™ CA Syringe Filters, Sterile, Globe Scientific
Filters Syringe Filters
Sterile Diamond® PureFlow™ syringe filters satisfy requirements of life science applications for sterilization of solutions or clarification of biological fluids. Sterile filters include a polypropylene (PP) microfiber pre-filter layer to improve filtration of particulate-laden fluids that are difficult to filter with single layer syringe filters.
  • CA (cellulose acetate) membrane
  • Low protein binding and biologically inert
  • Aqueous samples
  • High protein recovery from filtrate
  • Ideal for sample analysis that requires maximum recoveries, tissue culture media
More Product Information